accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 869901-13-3

IUPAC Name: 3-(chloromethyl)-1-methyl-5-phenyl-1H-pyrazole
Synonyms: 3-(chloromethyl)-1-methyl-5-phenylpyrazole
Molecular Weight (g/mol): 206.67
Molecular Formula: C11H11ClN2
InChi Key: KAZKAWDJSYXNGK-UHFFFAOYSA-N
SMILES: CN1N=C(CCl)C=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results