accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 869901-15-5

IUPAC Name: 1-methyl-5-(thiophen-2-yl)-1H-pyrazole-3-carboxylic acid
Synonyms: 1-methyl-5-(thiophen-2-yl)pyrazole-3-carboxylic acid
Molecular Weight (g/mol): 208.24
Molecular Formula: C9H8N2O2S
InChi Key: URCWVEMABYSGJA-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C1=CC=CS1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results