accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 869901-24-6

IUPAC Name: 4-[6-(tributylstannyl)pyridin-2-yl]morpholine
Synonyms: 4-[6-(tributylstannyl)pyridin-2-yl]morpholine
Molecular Weight (g/mol): 453.26
Molecular Formula: C21H38N2OSn
InChi Key: LDERBIPTTICDSG-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results