accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOO

CAS RN 87-22-9

IUPAC Name: 2-phenylethyl 2-hydroxybenzoate
Synonyms: phenethyl salicylate
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: YNMSDIQQNIRGDP-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)OCCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results