accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClClCl

CAS RN 87-40-1

IUPAC Name: 1,3,5-trichloro-2-methoxybenzene
Synonyms: tyrene C7H5cl3O
Molecular Weight (g/mol): 211.47
Molecular Formula: C7H5Cl3O
InChi Key: WCVOGSZTONGSQY-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)