accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87-59-2

CH3H2NH3C

CAS RN 87-59-2

IUPAC Name: 2,3-dimethylaniline
Synonyms: xylidine benzenamine, ar,ar-dimethyl-
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: VVAKEQGKZNKUSU-UHFFFAOYSA-N
SMILES: CC1=CC=CC(N)=C1C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)