accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClCl

CAS RN 87-61-6

IUPAC Name: 1,2,3-trichlorobenzene
Synonyms: 1,2,3-trichlorobenzene vic-trichlorobenzene
Molecular Weight (g/mol): 181.44
Molecular Formula: C6H3Cl3
InChi Key: RELMFMZEBKVZJC-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)