accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClCl

CAS RN 87-65-0

IUPAC Name: 2,6-dichlorophenol
Synonyms: 2,6-dichlorophenol 2,6-dichlorfenol rcra waste number U082
Molecular Weight (g/mol): 163
Molecular Formula: C6H4Cl2O
InChi Key: HOLHYSJJBXSLMV-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=CC=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (8)
Boiling Point 
  • (2)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (8)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)