CAS RN 87-66-1
Pyrogallol (Crystalline/Certified ACS), Fisher Chemical™
CAS: 87-66-1 Molecular Formula: C6H6O3 Molecular Weight (g/mol): 126.11 MDL Number: MFCD00002192 InChI Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N Synonym: pyrogallol,1,2,3-trihydroxybenzene,pyrogallic acid,1,2,3-benzenetriol,fouramine brown ap,fourrine pg,fourrine 85,pyro,c.i. oxidation base 32,piral PubChem CID: 1057 ChEBI: CHEBI:16164 IUPAC Name: benzene-1,2,3-triol SMILES: OC1=CC=CC(O)=C1O
Pyrogallol, 99%
CAS: 87-66-1 Molecular Formula: C6H6O3 Molecular Weight (g/mol): 126.11 MDL Number: MFCD00002192 InChI Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N Synonym: pyrogallol,1,2,3-trihydroxybenzene,pyrogallic acid,1,2,3-benzenetriol,fouramine brown ap,fourrine pg,fourrine 85,pyro,c.i. oxidation base 32,piral PubChem CID: 1057 ChEBI: CHEBI:16164 IUPAC Name: benzene-1,2,3-triol SMILES: OC1=CC=CC(O)=C1O
Pyrogallol, 98+%
CAS: 87-66-1 Molecular Formula: C6H6O3 Molecular Weight (g/mol): 126.11 MDL Number: MFCD00002192 InChI Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N Synonym: pyrogallol,1,2,3-trihydroxybenzene,pyrogallic acid,1,2,3-benzenetriol,fouramine brown ap,fourrine pg,fourrine 85,pyro,c.i. oxidation base 32,piral PubChem CID: 1057 ChEBI: CHEBI:16164 IUPAC Name: benzene-1,2,3-triol SMILES: OC1=CC=CC(O)=C1O
Pyrogallol, ACS, 99%
CAS: 87-66-1 Molecular Formula: C6H6O3 Molecular Weight (g/mol): 126.11 MDL Number: MFCD00002192 InChI Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N Synonym: pyrogallol,1,2,3-trihydroxybenzene,pyrogallic acid,1,2,3-benzenetriol,fouramine brown ap,fourrine pg,fourrine 85,pyro,c.i. oxidation base 32,piral PubChem CID: 1057 ChEBI: CHEBI:16164 IUPAC Name: benzene-1,2,3-triol SMILES: OC1=CC=CC(O)=C1O
Pyrogallic Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 87-66-1 Molecular Formula: C6H6O3 Molecular Weight (g/mol): 126.11 MDL Number: MFCD00002192 InChI Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N Synonym: pyrogallol,1,2,3-trihydroxybenzene,pyrogallic acid,1,2,3-benzenetriol,fouramine brown ap,fourrine pg,fourrine 85,pyro,c.i. oxidation base 32,piral PubChem CID: 1057 ChEBI: CHEBI:16164 IUPAC Name: benzene-1,2,3-triol SMILES: OC1=CC=CC(O)=C1O
Pyrogallol, ACS reagent
CAS: 87-66-1 Molecular Formula: C6H6O3 Molecular Weight (g/mol): 126.11 MDL Number: MFCD00002192 InChI Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N Synonym: pyrogallol,1,2,3-trihydroxybenzene,pyrogallic acid,1,2,3-benzenetriol,fouramine brown ap,fourrine pg,fourrine 85,pyro,c.i. oxidation base 32,piral PubChem CID: 1057 ChEBI: CHEBI:16164 IUPAC Name: benzene-1,2,3-triol SMILES: OC1=CC=CC(O)=C1O
phyproof™ Pyrogallol, ≥98% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Pyrogallol, Crystal, Reagent, ACS, Spectrum™ Chemical
CAS: 87-66-1 Molecular Formula: C6H6O3 Molecular Weight (g/mol): 126.11 MDL Number: MFCD00002192 InChI Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N IUPAC Name: benzene-1,2,3-triol SMILES: OC1=CC=CC(O)=C1O
Pyrogallol, Crystal, Reagent, ACS, Spectrum™ Chemical
CAS: 87-66-1 Molecular Formula: C6H6O3 Molecular Weight (g/mol): 126.11 MDL Number: MFCD00002192 InChI Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N IUPAC Name: benzene-1,2,3-triol SMILES: OC1=CC=CC(O)=C1O
Pyrogallol 99.0+%, TCI America™
CAS: 87-66-1 Molecular Formula: C6H6O3 Molecular Weight (g/mol): 126.11 MDL Number: MFCD00002192 InChI Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N Synonym: pyrogallol,1,2,3-trihydroxybenzene,pyrogallic acid,1,2,3-benzenetriol,fouramine brown ap,fourrine pg,fourrine 85,pyro,c.i. oxidation base 32,piral PubChem CID: 1057 ChEBI: CHEBI:16164 IUPAC Name: benzene-1,2,3-triol SMILES: OC1=CC=CC(O)=C1O
Alkaline Pyrogallol Reagent, Orsat, Ricca Chemical
CAS: 87-66-1 Molecular Formula: C6H6O3 Molecular Weight (g/mol): 126.11 MDL Number: MFCD00002192 InChI Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N Synonym: pyrogallol,1,2,3-trihydroxybenzene,pyrogallic acid,1,2,3-benzenetriol,fouramine brown ap,fourrine pg,fourrine 85,pyro,c.i. oxidation base 32,piral PubChem CID: 1057 ChEBI: CHEBI:16164 IUPAC Name: benzene-1,2,3-triol SMILES: OC1=CC=CC(O)=C1O
Sigma Aldrich Pyrogallol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Fisher Science Education™ Pyrogallic Acid
CAS: 87-66-1 Molecular Formula: C6H6O3 Molecular Weight (g/mol): 126.11 MDL Number: MFCD00002192 InChI Key: WQGWDDDVZFFDIG-UHFFFAOYSA-N Synonym: pyrogallol,1,2,3-trihydroxybenzene,pyrogallic acid,1,2,3-benzenetriol,fouramine brown ap,fourrine pg,fourrine 85,pyro,c.i. oxidation base 32,piral PubChem CID: 1057 ChEBI: CHEBI:16164 IUPAC Name: benzene-1,2,3-triol SMILES: OC1=CC=CC(O)=C1O