accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNONClONClO

CAS RN 87-90-1

IUPAC Name: trichloro-1,3,5-triazinane-2,4,6-trione
Synonyms: trichloroisocyanuric acid symclosene trichloroiminocyanuric acid trichlorocyanuric acid
Molecular Weight (g/mol): 232.4
Molecular Formula: C3Cl3N3O3
InChi Key: YRIZYWQGELRKNT-UHFFFAOYSA-N
SMILES: ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)