accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 870-08-6

IUPAC Name: dioctylstannanone
Synonyms: stannane, dioctyloxo-
Molecular Weight (g/mol): 361.16
Molecular Formula: C16H34OSn
InChi Key: LQRUPWUPINJLMU-UHFFFAOYSA-N
SMILES: CCCCCCCC[Sn](=O)CCCCCCCC
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)