accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH3Cl

CAS RN 870-24-6

IUPAC Name: 2-chloroethan-1-aminium chloride
Synonyms: 2-chloroethanaminium chloride
Molecular Weight (g/mol): 115.99
Molecular Formula: C2H7Cl2N
InChi Key: ONRREFWJTRBDRA-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]CCCl
Percent Purity 
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)