accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2OOHCl

CAS RN 870621-17-3

IUPAC Name: benzyl 3-aminopyrrolidine-1-carboxylate hydrochloride
Synonyms: benzyl 3-aminopyrrolidine-1-carboxylate hydrochloride
Molecular Weight (g/mol): 256.73
Molecular Formula: C12H17ClN2O2
InChi Key: QNQVBYGRFHOBNO-UHFFFAOYNA-N
SMILES: Cl.NC1CCN(C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)