accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FSOOCl

CAS RN 870704-14-6

IUPAC Name: 2-fluoro-5-methylbenzene-1-sulfonyl chloride
Synonyms: 2-fluoro-5-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 208.63
Molecular Formula: C7H6ClFO2S
InChi Key: MOZTWWZAQLPJMT-UHFFFAOYSA-N
SMILES: CC1=CC(=C(F)C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results