CAS RN 871-70-5
CAS RN 871-70-5
IUPAC Name:
octadecanedioic acid
Synonyms:
octadecanedioic acid
Molecular Weight (g/mol):
314.47
Molecular Formula:
C18H34O4
InChi Key:
BNJOQKFENDDGSC-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCCCCCCCCCCCCCC(O)=O
1
–
1
of
1
results
1
Octadecanedioic Acid 98.0+%, TCI America™
CAS: 871-70-5 Molecular Formula: C18H34O4 Molecular Weight (g/mol): 314.466 MDL Number: MFCD00142369 InChI Key: BNJOQKFENDDGSC-UHFFFAOYSA-N Synonym: 1,16-hexadecanedicarboxylic acid,1,18-octadecanedioic acid,unii-rsz6pq0qqj,octadecane-1,18-dioic acid,rsz6pq0qqj,hexadecanedicarboxylic acid,1,18-octadecadioic acid,octadecanedioate,1,18-octadecanedioate,octadecandis PubChem CID: 70095 IUPAC Name: octadecanedioic acid SMILES: C(CCCCCCCCC(=O)O)CCCCCCCC(=O)O