accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHHNCH3O

CAS RN 871-78-3

IUPAC Name: N-(2-acetamidoethyl)acetamide
Synonyms: N-(2-acetamidoethyl)acetamide
Molecular Weight (g/mol): 144.17
Molecular Formula: C6H12N2O2
InChi Key: WNYIBZHOMJZDKN-UHFFFAOYSA-N
SMILES: CC(=O)NCCNC(C)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)