accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPCH3CH3O

CAS RN 871-95-4

IUPAC Name: 1-(dimethylphosphoryl)dodecane
Synonyms: 1-(dimethylphosphoryl)dodecane
Molecular Weight (g/mol): 246.38
Molecular Formula: C14H31OP
InChi Key: SIDULKZCBGMXJL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCP(C)(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results