accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87100-28-5

IUPAC Name: 2-benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 218.1
Molecular Formula: C13H19BO2
InChi Key: YCNQPAVKQPLZRS-UHFFFAOYSA-N
SMILES: CC1(C)OB(CC2=CC=CC=C2)OC1(C)C
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)