accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFOHOOH

CAS RN 871126-15-7

IUPAC Name: (3-fluoro-2-formylphenyl)boronic acid
Synonyms: 3-fluoro-2-formylphenylboronic acid
Molecular Weight (g/mol): 167.93
Molecular Formula: C7H6BFO3
InChi Key: GVHWLCYABLWGIR-UHFFFAOYSA-N
SMILES: OB(O)C1=C(C=O)C(F)=CC=C1
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)