accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87120-72-7

IUPAC Name: 1-[(tert-butoxy)carbonyl]piperidin-4-aminium
Synonyms: 1-(tert-butoxycarbonyl)piperidin-4-aminium
Molecular Weight (g/mol): 201.29
Molecular Formula: C10H21N2O2
InChi Key: LZRDHSFPLUWYAX-UHFFFAOYSA-O
SMILES: CC(C)(C)OC(=O)N1CCC([NH3+])CC1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (4)
  • (4)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (6)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)