CAS RN 871329-52-1
IUPAC Name:
(2-chloro-3-fluorophenyl)boronic acid
Synonyms:
2-chloro-3-fluorophenylboronic acid
Molecular Weight (g/mol):
174.36
Molecular Formula:
C6H5BClFO2
InChi Key:
TYOGIUGNZOBBHE-UHFFFAOYSA-N
SMILES:
OB(O)C1=C(Cl)C(F)=CC=C1
1
–
1
of
1
results
1
Promotions Available
2-Chloro-3-fluorophenylboronic acid, 97%
2-Chloro-3-fluorophenylboronic acid, 97%
CAS: 871329-52-1 Molecular Formula: C6H5BClFO2 Molecular Weight (g/mol): 174.36 MDL Number: MFCD07363741 InChI Key: TYOGIUGNZOBBHE-UHFFFAOYSA-N Synonym: 2-chloro-3-fluorophenyl boronic acid,1-borono-2-chloro-3-fluorobenzene,2-chloro-3-fluorobenzeneboronic acid,pubchem19756,acmc-209qfx,2-chloro-3-fluoro-phenylboronic acid,2-chloro-3-fluoro-benzeneboronic acid,2-chloro-3-fluoro-phenyl boronic acid,2-chloro-3-fluorophenyl ;boronic acid PubChem CID: 22309459 IUPAC Name: (2-chloro-3-fluorophenyl)boronic acid SMILES: OB(O)C1=C(Cl)C(F)=CC=C1