accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH3CCH3HOONa

CAS RN 871329-70-3

IUPAC Name: sodium hydrogen (4,4-dimethylcyclohexa-1,5-dien-1-yl)boronate
Synonyms: sodium hydrogen 4,4-dimethylcyclohexa-1,5-dien-1-ylboronate
Molecular Weight (g/mol): 173.98
Molecular Formula: C8H12BNaO2
InChi Key: YBUXCAFVTNSKLJ-UHFFFAOYSA-N
SMILES: [Na+].CC1(C)CC=C(C=C1)B(O)[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results