accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 871329-71-4

IUPAC Name: (5-oxocyclopent-1-en-1-yl)boronic acid
Synonyms: 5-oxocyclopent-1-en-1-ylboronic acid
Molecular Weight (g/mol): 125.92
Molecular Formula: C5H7BO3
InChi Key: LEMUMNFZUFSWGV-UHFFFAOYSA-N
SMILES: OB(O)C1=CCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results