accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNSOOH

CAS RN 871825-56-8

IUPAC Name: 1-methyl-3-(thiophen-2-yl)-1H-pyrazole-5-carboxylic acid
Synonyms: 2-methyl-5-(thiophen-2-yl)pyrazole-3-carboxylic acid
Molecular Weight (g/mol): 208.24
Molecular Formula: C9H8N2O2S
InChi Key: YSTOGMCNCZGBTG-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C(O)=O)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results