accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 871940-31-7

IUPAC Name: (3-cyano-2,4-difluorophenyl)boronic acid
Synonyms: 3-cyano-2,4-difluorophenylboronic acid
Molecular Weight (g/mol): 182.92
Molecular Formula: C7H4BF2NO2
InChi Key: VQEKFJVJHSVFTM-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C(C#N)=C(F)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results