accessibility menu, dialog, popup

UserName

Viewing profile as user:
BHOHOOH

CAS RN 87199-14-2

IUPAC Name: [2-(hydroxymethyl)phenyl]boronic acid
Synonyms: 2-(hydroxymethyl)phenylboronic acid
Molecular Weight (g/mol): 151.96
Molecular Formula: C7H9BO3
InChi Key: AFHOBSCDNXGFMO-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results