accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrS

CAS RN 872-31-1

IUPAC Name: 3-bromothiophene
Synonyms: 3-bromothiophene
Molecular Weight (g/mol): 163.03
Molecular Formula: C4H3BrS
InChi Key: XCMISAPCWHTVNG-UHFFFAOYSA-N
SMILES: BrC1=CSC=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)