accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CHN

CAS RN 872-32-2

IUPAC Name: 2-methylpyrrolidin-2-ylium
Synonyms: 2-methylpyrrolidin-2-ylium
Molecular Weight (g/mol): 84.14
Molecular Formula: C5H10N
InChi Key: OZLJZWIVBYLSOK-UHFFFAOYSA-N
SMILES: C[C+]1CCCN1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)