accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHSH

CAS RN 872-35-5

IUPAC Name: 1H-imidazole-2-thiol
Synonyms: 2-mercaptoimidazole
Molecular Weight (g/mol): 100.14
Molecular Formula: C3H4N2S
InChi Key: OXFSTTJBVAAALW-UHFFFAOYSA-N
SMILES: SC1=NC=CN1
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)