CAS RN 872460-12-3
IUPAC Name:
5-(dihydroxyboranyl)-2-fluorobenzoic acid
Synonyms:
5-(dihydroxyboranyl)-2-fluorobenzoic acid
Molecular Weight (g/mol):
183.93
Molecular Formula:
C7H6BFO4
InChi Key:
DGORTXQWDDXSIQ-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(C(O)=O)=C(F)C=C1