accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 872679-70-4

IUPAC Name: 3-{2-[2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)ethoxy]ethoxy}propanoic acid
Synonyms: 3-[2-(2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}ethoxy)ethoxy]propanoic acid
Molecular Weight (g/mol): 399.44
Molecular Formula: C22H25NO6
InChi Key: QWHLFJJLRVOHTM-UHFFFAOYSA-N
SMILES: OC(=O)CCOCCOCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)