CAS RN 872726-44-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ NSC 721648
A cell-permeable benzothiazole compound that exhibits potent in vitro antiproliferative activity against HCC 2998, IGROV1, MCF-7, MDA 468, SR, and T-47D.