accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 873-38-1

IUPAC Name: 2-bromo-4-chloroaniline
Synonyms: 2-bromo-4-chloroaniline
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: SYTBIFURTZACKR-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1Br
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)