accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSO

CAS RN 873-55-2

IUPAC Name: benzenesulfinate
Synonyms: benzenesulfinate
Molecular Weight (g/mol): 141.16
Molecular Formula: C6H5O2S
InChi Key: JEHKKBHWRAXMCH-UHFFFAOYSA-M
SMILES: [O-]S(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)