accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNN(E)

CAS RN 873-69-8

IUPAC Name: (E)-N-[(pyridin-2-yl)methylidene]hydroxylamine
Synonyms: (E)-pyridine-2-aldoxime
Molecular Weight (g/mol): 122.13
Molecular Formula: C6H6N2O
InChi Key: MTFJSAGADRTKCI-VMPITWQZSA-N
SMILES: O\N=C\C1=NC=CC=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results