accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 873-74-5

IUPAC Name: 4-aminobenzonitrile
Synonyms: P-aminobenzonitrile
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: YBAZINRZQSAIAY-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (4)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (5)
  • (5)
  • (2)
  • (8)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)