accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 873-75-6

IUPAC Name: (4-bromophenyl)methanol
Synonyms: para-bromobenzyl alcohol C7H7BrO
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: VEDDBHYQWFOITD-UHFFFAOYSA-N
SMILES: OCC1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)