accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 873-77-8

IUPAC Name: bromo(4-chlorophenyl)magnesium
Synonyms: bromo(4-chlorophenyl)magnesium
Molecular Weight (g/mol): 215.76
Molecular Formula: C6H4BrClMg
InChi Key: CDEMHJCJMMOFMB-UHFFFAOYSA-M
SMILES: ClC1=CC=C([Mg]Br)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)