accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 873-94-9

IUPAC Name: 3,3,5-trimethylcyclohexan-1-one
Synonyms: 3,3,5-trimethylcyclohexan-1-one
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: POSWICCRDBKBMH-UHFFFAOYNA-N
SMILES: CC1CC(=O)CC(C)(C)C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)