accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87314-12-3

IUPAC Name: λ³-nickel(3+) bis((3,4,6-trichloro-2-sulfanidylphenyl)sulfanide) tetrabutylazanium
Synonyms: λ³-nickel(3+) bis((3,4,6-trichloro-2-sulfanidylphenyl)sulfanide) tetrabutylammonium ion
Molecular Weight (g/mol): 788.25
Molecular Formula: C28H38Cl6NNiS4
InChi Key: WIBIOFAUOGMVAE-UHFFFAOYSA-J
SMILES: [Ni+3].[S-]C1=C([S-])C(Cl)=C(Cl)C=C1Cl.[S-]C1=C([S-])C(Cl)=C(Cl)C=C1Cl.CCCC[N+](CCCC)(CCCC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results