CAS RN 87327-69-3
IUPAC Name:
1-(2-chloro-6-fluorophenyl)ethan-1-one
Synonyms:
1-(2-chloro-6-fluorophenyl)ethanone
Molecular Weight (g/mol):
172.58
Molecular Formula:
C8H6ClFO
InChi Key:
DNVGZKIRMBCQEQ-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(F)C=CC=C1Cl
1
–
1
of
1
results
1
2'-Chloro-6'-fluoroacetophenone, 97%, Thermo Scientific™
CAS: 87327-69-3 Molecular Formula: C8H6ClFO Molecular Weight (g/mol): 172.58 MDL Number: MFCD01631390 InChI Key: DNVGZKIRMBCQEQ-UHFFFAOYSA-N PubChem CID: 2773578 SMILES: CC(=O)C1=C(F)C=CC=C1Cl