CAS RN 87333-19-5
IUPAC Name:
(2S,3aS,6aS)-1-[(2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]-octahydrocyclopenta[b]pyrrole-2-carboxylic acid
Synonyms:
ramipril|altace|cardace|tritace|carasel|delix|triatec|ramace|vesdil|acovil
Molecular Weight (g/mol):
416.52
Molecular Formula:
C23H32N2O5
InChi Key:
HDACQVRGBOVJII-JBDAPHQKSA-N
SMILES:
CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1[C@H]2CCC[C@H]2C[C@H]1C(O)=O
1
–
2
of
2
results
2
Ramipril Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Ramipril is an angiotensin-converting-enzyme (ACE) inhibitor, which acts on the renin angiotensin aldosterone system. It is widely used in reducing hypertension by the suppression of the effects of angiotensin II at its receptor, thereby blocking the renin-angiotensin system.