accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 873431-46-0

IUPAC Name: 4-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Synonyms: 4-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Molecular Weight (g/mol): 307.17
Molecular Formula: C16H23BFNO3
InChi Key: YOJYRVSDQHWBGS-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(F)=C(C=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results