accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONI

CAS RN 87350-77-4

IUPAC Name: 4-(4-iodophenyl)morpholine
Synonyms: 4-(4-iodophenyl)morpholine
Molecular Weight (g/mol): 289.12
Molecular Formula: C10H12INO
InChi Key: YUEREUIIUISPFV-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)N1CCOCC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)