accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 873985-57-0

IUPAC Name: 3-nitro-N-(2-nitrophenyl)benzene-1-sulfonamide
Synonyms: 3-nitro-N-(2-nitrophenyl)benzenesulfonamide
Molecular Weight (g/mol): 323.28
Molecular Formula: C12H9N3O6S
InChi Key: ODFYBQONMXQKNR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1)S(=O)(=O)NC1=CC=CC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results