accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874-05-5

IUPAC Name: 1H-indazol-3-amine
Synonyms: 3-aminoindazole
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: YDTDKKULPWTHRV-UHFFFAOYSA-N
SMILES: NC1=NNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results