accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NSN

CAS RN 874-37-3

IUPAC Name: 2,1,3-benzothiadiazol-5-amine
Synonyms: 2,1,3-benzothiadiazol-5-amine
Molecular Weight (g/mol): 151.19
Molecular Formula: C6H5N3S
InChi Key: JRJPKRSKPOCUEV-UHFFFAOYSA-N
SMILES: NC1=CC2=NSN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results