accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOCl

CAS RN 874-42-0

IUPAC Name: 2,4-dichlorobenzaldehyde
Synonyms: 2,4-dichlorobenzaldehyde
Molecular Weight (g/mol): 175.01
Molecular Formula: C7H4Cl2O
InChi Key: YSFBEAASFUWWHU-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=O)C(Cl)=C1
Boiling Point 
  • (8)
  • (1)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (6)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (8)
  • (1)
  • (5)
  • (4)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)