accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClO

CAS RN 874-60-2

IUPAC Name: 4-methylbenzoyl chloride
Synonyms: benzoyl chloride, 4-methyl-
Molecular Weight (g/mol): 154.59
Molecular Formula: C8H7ClO
InChi Key: NQUVCRCCRXRJCK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (6)
  • (4)
Percent Purity 
  • (3)
  • (7)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (7)
  • (3)
  • (3)
  • (3)
  • (4)
  • (3)
  • (6)
  • (4)
  • (3)
  • (7)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (10)