accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOO

CAS RN 874-61-3

IUPAC Name: 4-oxocyclohexane-1-carboxylic acid
Synonyms: 4-oxocyclohexanecarboxylate
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: OWLXUYGCLDGHJJ-UHFFFAOYSA-N
SMILES: OC(=O)C1CCC(=O)CC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)